Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Neuroglycan C
- Antigen Symbol:CSPG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10675
- Antigen Synonyms:Neuroglycan C|NGCAcidic leucine-rich EGF-like domain-containing brain protein|MGC44034|chondroitin sulfate proteoglycan 5|CALEB|chondroitin sulfate proteoglycan 5 (neuroglycan C)
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to CSPG5(chondroitin sulfate proteoglycan 5 (neuroglycan C)) The peptide sequence was selected from the N terminal of CSPG5. Peptide sequence LGGVTAEAGSGDAQALPATLQAPHEVLGQSIMPPAIPEATEASGPPSPTP.
- Purification:Immunogen affinity purified
- Cat. No.:102192-378
- Supplier no.:NBP1-59341
The Neuroglycan C / CSPG5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Neuroglycan C / CSPG5. This antibody reacts with human. The Neuroglycan C / CSPG5 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CSPG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human