Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:MAP1S
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:55201
- Antigen Synonyms:C19orf5|VCY2 interacting protein 1|MAP8MAP-1S|Microtubule-associated protein 8|FLJ10669|Variable charge Y chromosome 2-interacting protein 1|microtubule-associated protein 1S|VCY2IP1BPY2-interacting protein 1|chromosome 19 open reading frame 5|BPY2 interacting protein 1|VCY2-interacting protein 1|BPY2IP1VCY2IP-1|MGC133087
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 525 and 575 of human MAP1S using the numbering given in entry NP_060644.4 (GeneID 55201).
- Purification:Immunogen affinity purified
- Cat. No.:102176-978
- Supplier no.:NBP1-47282
Rabbit Polyclonal MAP8 Antibody. Tested Applications: Immunoprecipitation, Western Blot (-). Tested Reactivity: Human.
Type: Primary
Antigen: MAP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human