Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Leucine Rich Repeat Containing 56
- Antigen Symbol:LRRC56
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:115399
- Antigen Synonyms:leucine rich repeat containing 56
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to LRRC56(leucine rich repeat containing 56) The peptide sequence was selected from the N terminal of LRRC56. Peptide sequence LEMCVDTREGSLGNFGVHLPNLDQLKLNGSHLGSLRDLGTSLGHLQVLWL.
- Purification:Immunogen affinity purified
- Cat. No.:102199-864
- Supplier no.:NBP1-70626
Specifications
About this item
Rabbit Polyclonal LRRC56 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.
Type: Primary
Antigen: LRRC56
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human