Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon beta
- Antigen Symbol:IFNB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:3456
- Antigen Synonyms:IFBIFNB|interferon beta|interferon|MGC96956|beta 1|Fibroblast interferon|IFF|IFN-beta|fibroblast
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 17 amino acid synthetic peptide from near the center of human IFN-b. The immunogen is located within amino acids 110 - 160 of IFN-beta.
- Purification:Immunogen affinity purified
- Cat. No.:102211-326
- Supplier no.:NBP1-77288
Specifications
About this item
Rabbit Polyclonal Interferon beta Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml
Type: Primary
Antigen: Interferon beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human