To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GW182
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:27327
- Antigen Synonyms:CAGH26FLJ22043|GW182 autoantigen|KIAA1460EDIE|DKFZp666E117|trinucleotide repeat-containing gene 6A protein|trinucleotide repeat containing 6|TNRC6|Glycine-tryptophan protein of 182 kDa|trinucleotide repeat containing 6A|EMSY interactor protein|GW182GW1|MGC75384|CAG repeat protein 26|Protein GW1
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1225 and 1275 of human glycine-tryptophan protein of 182 kDa (GW182) using the numbering given in entry NP_055309.2 (GeneID 27327).
- Purification:Immunogen affinity purified
- Cat. No.:102162-552
- Supplier no.:NBP1-28702
Specifications
About this item
Rabbit Polyclonal GW182 Antibody. Tested Applications: Immunoprecipitation, Western Blot (-). Tested Reactivity: Human.
Type: Primary
Antigen: GW182
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human