To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:G-protein coupled Receptor 120
- Antigen Symbol:FFAR4/GPR120
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:338557
- Antigen Synonyms:G-protein coupled receptor PGR4|G-protein coupled receptor GT01|GPR120|G protein-coupled receptor 120|PGR4DKFZp686F0824|GT01|G-protein-coupled receptor GT01|G-protein coupled receptor 120|G protein-coupled receptor 129|MGC119984|G protein-coupled receptor PGR4|omega-3 fatty acid receptor 1|G-protein coupled receptor 129|GPR129
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:A synthetic peptide made to an internal portion of the human GPR120 protein (between residues 200-300) [UniProt Q5NUL3]
- Purification:Immunogen affinity purified
- Cat. No.:102154-950
- Supplier no.:NBP1-00858
Specifications
About this item
The FFAR4 / GPR120 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FFAR4 / GPR120. This antibody reacts with human, mouse, rat. The FFAR4 / GPR120 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FFAR4/GPR120
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat