Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Connexin 40
- Antigen Symbol:Cx40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Form:Lyophilized
- Gene ID:2702
- Antigen Synonyms:MGC11185|40kD (connexin 40)|alpha 5|connexin-40|gap junction protein|gap junction alpha-5 protein|40kDa (connexin 40)|40kDa|Cx40
- Storage Buffer:Whole antisera
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide from 2d cytoplasmic domain of human 2d cytoplasmic domain of Connexin-40 (CX40, CXA5) conjugated to an immunogenic carrier protein was used as the antigen.
- Purification:Unpurified
- Cat. No.:102176-372
- Supplier no.:NBP1-46568
Specifications
About this item
The Connexin 40 / GJA5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Connexin 40 / GJA5. This antibody reacts with mouse, rat. The Connexin 40 / GJA5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Cx40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat