Anti-eIF2B epsilon Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The eIF2B epsilon Antibody from Novus Biologicals is a rabbit polyclonal antibody to eIF2B epsilon. This antibody reacts with human. The eIF2B epsilon Antibody has been validated for the following applications: Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Type: Primary
Antigen: eIF2B epsilon
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
NBP1-30449.pdf
Datasheet
Specifications
- Size:100 μl
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:Immunogen affinity purified
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Isotype:IgG
- Host:Rabbit
- Concentration:1.0 mg/ml
- Cat. No.:102163-508
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:eIF2B epsilon
- Gene ID:8893
- Immunogen:The immunogen for this product maps to a region between residue 550 and 600 of human eukaryotic translation initiation factor 2B, subunit 5 epsilon using the numbering given in entry NP_003898.2 (GeneID 8893).
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:subunit 5 (epsilon|LVWM|82kD)|subunit 5 epsilon|eukaryotic translation initiation factor 2B|EIF2BE|82kDa|EIF-2B|translation initiation factor eIF-2B subunit epsilon|CLE|CACH|eIF-2B GDP-GTP exchange factor subunit epsilon|EIF2Bepsilon
- Antigen Name:EIF2B epsilon
- Storage Temperature:Store at 4C. Do not freeze.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



