Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DPP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:0.05 mg
- Gene ID:57628
- Antigen Synonyms:Dipeptidyl peptidase X|DPPY|dipeptidyl-peptidase 10 (non-functional)|inactive dipeptidyl peptidase 10|dipeptidyl peptidase like protein 2|DPP X|dipeptidyl peptidase 10|KIAA1492|DPRP-3|DPL2|DPRP3dipeptidylpeptidase 10|Dipeptidyl peptidase-like protein 2|dipeptidyl-peptidase 10|Dipeptidyl peptidase IV-related protein 3
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 17 amino acid peptide from internal region of human DPP10.
- Purification:Immunogen affinity purified
- Cat. No.:102173-942
- Supplier no.:NBP1-44338
Specifications
About this item
The DPP10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DPP10. This antibody reacts with human. The DPP10 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DPP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human