Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:DEAD (Asp-Glu-Ala-Asp) box helicase 24
- Antigen Symbol:DDX24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:57062
- Antigen Synonyms:DEAD (Asp-Glu-Ala-Asp) box polypeptide 24|ATP-dependent RNA helicase DDX24|EC 3.6.4.13|DEAD box protein 24|S. cerevisiae CHL1-like helicase|EC 3.6.1|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 using the numbering given in entry NP_065147.1 (GeneID 57062).
- Purification:Immunogen affinity purified
- Cat. No.:102106-872
- Supplier no.:NB100-2224
The DDX24 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DDX24. This antibody reacts with human. The DDX24 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: DDX24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human