Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Discoidin Domain receptor tyrosine kinase 1
- Antigen Symbol:DDR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Primates,Bat,Pig,Horse,Monkey,Human,Bovine,Dog
- Size:0.05 mg
- Gene ID:780
- Antigen Synonyms:HGK2|EC 2.7.10|MCK10|type 4|CD167 antigen-like family member A|Protein-tyrosine kinase 3A|receptor|EC 2.7.10.1|Epithelial discoidin domain receptor 1|NTRK4|EDDR1|MCK-10|PTK3A protein tyrosine kinase 3A|ENTRK4|member 1|discoidin domain receptor family|discoidin domain receptor tyrosine kinase 1|Discoidin receptor tyrosine kinase|CD167a antigen|PTK3A|TRK E|DDR
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 16 amino acid peptide from C-terminus of human DDR1.
- Purification:Immunogen affinity purified
- Cat. No.:102121-788
- Supplier no.:NB110-39011
Specifications
About this item
The DDR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DDR1. This antibody reacts with human, porcine, bat, bovine, canine, equine, monkey, primate. The DDR1 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 24 ?g/ml
Type: Primary
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Porcine, Bat, Bovine, Canine, Equine, Monkey, Primate