Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-X-C Motif) Receptor 4
- Antigen Symbol:CXCR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Primates,Hamster,Rabbit,Monkey,Chicken,Human,Rat,Mouse
- Size:0.05 ml
- Gene ID:7852
- Antigen Synonyms:C-X-C chemokine receptor type 4|leukocyte-derived seven-transmembrane-domain receptor|Leukocyte-derived seven transmembrane domain receptor|NPYR|HM89NPYRL|neuropeptide Y receptor Y3|seven-transmembrane-segment receptor|CXCR-4|seven transmembrane helix receptor|chemokine (C-X-C motif) receptor 4|CXC-R4|D2S201ESDF-1 receptor|lipopolysaccharide-associated protein 3|receptor 4 (fusin)|chemokine (C-X-C motif)|NPY3RWHIM|NPYY3R|LCR1|Fusin|HSY3RR|LESTRCD184 antigen|CD184|spleen|LAP3|FB22|Stromal cell-derived factor 1 receptor
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human CXCR4.
- Purification:Immunogen affinity purified
- Cat. No.:102253-092
- Supplier no.:NLS1380
Specifications
About this item
The CXCR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CXCR4. This antibody reacts with human, mouse, rat, chicken, hamster, monkey, primate, rabbit. The CXCR4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Hamster, Monkey, Primate, Rabbit