Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Cleavage And polyadenylation specific Factor 6 68kDa
- Antigen Symbol:CPSF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:11052
- Antigen Synonyms:pre-mRNA cleavage factor I|68kD subunit|cleavage and polyadenylation specific factor 6|Pre-mRNA cleavage factor Im 68 kDa subunit|pre-mRNA cleavage factor Im (68kD)|CFIM|HPBRII-7|CFIM68CPSF 68 kDa subunit|CFIMcleavage and polyadenylation specific factor 6|HPBRII-4|68kDa|CFIm68|Cleavage and polyadenylation specificity factor 68 kDa subunit|Protein HPBRII-4/7|cleavage and polyadenylation specificity factor subunit 6
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cleavage and polyadenylation specific factor 6, 68kDa using the numbering given in entry NP_008938.1 (GeneID 11052).
- Purification:Immunogen affinity purified
- Cat. No.:102111-250
- Supplier no.:NB100-61596
Specifications
About this item
The CPSF6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CPSF6. This antibody reacts with human, mouse. The CPSF6 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CPSF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse