About this item
The Collagen I alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Collagen I alpha 1. This antibody reacts with human, mouse. The Collagen I alpha 1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: Collagen I alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102211-760
- 102211-758
- Antigen Symbol:
- COL1
- COL1
- Antigen Name:
- Collagen I alpha 1
- Collagen I alpha 1
- Antigen Synonyms:
- Alpha-1 type I collagen|alpha 1|Collagen 1|OI4|Collagen1|collagen of skin|collagen alpha-1(I) chain|collagen|pro-alpha-1 collagen type 1|Collagen 1 alpha 1|Collagen-1|alpha-1 chain|collagen alpha 1 chain type I|tendon and bone|type I
- Alpha-1 type I collagen|alpha 1|Collagen 1|OI4|Collagen1|collagen of skin|collagen alpha-1(I) chain|collagen|pro-alpha-1 collagen type 1|Collagen 1 alpha 1|Collagen-1|alpha-1 chain|collagen alpha 1 chain type I|tendon and bone|type I
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1277
- 1277
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to an internal portion of the human Collagen I protein (between residues 150-200) [UniProt P02452]
- A synthetic peptide made to an internal portion of the human Collagen I protein (between residues 150-200) [UniProt P02452]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1.1 mg/ml
- 1.1 mg/ml
- Storage Buffer:
- PBS and 30% Glycerol
- PBS and 30% Glycerol
- Storage Temperature:
- Aliquot and store at −20 °C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at −20 °C or -80C. Avoid freeze-thaw cycles.