To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CL-P1/COLEC12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10978
- Antigen Synonyms:Pre-mRNA cleavage complex II protein Clp1|CLP1|HEABATP/GTP-binding protein|homolog (S. cerevisiae)|hClp1|EC 2.7.1.78|homolog of yeast CFIA subunit Clp1p|Polynucleotide kinase Clp1|polyribonucleotide 5'-hydroxyl-kinase Clp1|cleavage and polyadenylation factor I subunit
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:The immunogen for this antibody is CLP1 - C-terminal region. Peptide sequence KVGAPTIPDSCLPLGMSQEDNQLKLVPVTPGRDMVHHLLSVSTAEGTEEN.
- Purification:Immunogen affinity purified
- Cat. No.:102223-220
- Supplier no.:NBP1-98452
The CL-P1 / COLEC12 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CL-P1 / COLEC12. This antibody reacts with human. The CL-P1 / COLEC12 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CL-P1/COLEC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human