Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Chromatin Assembly Factor 1, Subunit B (p60)
- Antigen Symbol:CHAF1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:8208
- Antigen Synonyms:chromatin assembly factor 1|CAF-1|CAF1P60CAF-I 60 kDa subunit|chromatin assembly factor 1 subunit B|MPHOSPH7CAF-I p60|CAF1ACAF-1 subunit B|subunit B (p60)|human chromatin assembly factor-I p60 subunit|CAF-IP60|Chromatin assembly factor I p60 subunit|MPP7M-phase phosphoprotein 7|10|CAF1
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 509 and the C-terminus (residue 559) of human chromatin assembly factor 1, subunit B (p60) using the numbering given in entry NP_005432.1 (GeneID 8208).
- Purification:Immunogen affinity purified
- Cat. No.:102110-484
- Supplier no.:NB100-57523
Specifications
About this item
The CHAF1B Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHAF1B. This antibody reacts with human. The CHAF1B Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CHAF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human