About this item
The CCR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCR2. This antibody reacts with human, mouse. The CCR2 Antibody has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102178-554
- 102178-568
- Antigen Symbol:
- CCR2
- CCR2
- Antigen Name:
- Chemokine (C-C motif) receptor 2
- Chemokine (C-C motif) receptor 2
- Antigen Synonyms:
- MCP-1 receptor|C-C chemokine receptor type 2|CD192 antigen|CKR2|FLJ78302|C-C CKR-2|CCR-2|CMKBR2MGC111760|CKR2A|Monocyte chemoattractant protein 1 receptor|MGC168006|CCR2A|monocyte chemotactic protein 1 receptor|MCP-1-RMGC103828|CC-CKR-2CKR2B|CD192|CCR2B|chemokine (C-C motif) receptor 2
- MCP-1 receptor|C-C chemokine receptor type 2|CD192 antigen|CKR2|FLJ78302|C-C CKR-2|CCR-2|CMKBR2MGC111760|CKR2A|Monocyte chemoattractant protein 1 receptor|MGC168006|CCR2A|monocyte chemotactic protein 1 receptor|MCP-1-RMGC103828|CC-CKR-2CKR2B|CD192|CCR2B|chemokine (C-C motif) receptor 2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 729230
- 729230
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide made to an N-terminal portion of the mouse CCR2 protein (within residues 20-100). [Swiss-Prot# P51683]
- Synthetic peptide made to an N-terminal portion of the mouse CCR2 protein (within residues 20-100). [Swiss-Prot# P51683]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- PBS and 30% Glycerol
- PBS and 30% Glycerol
- Storage Temperature:
- Store at -20C. Avoid freeze-thaw cycles.
- Store at -20C. Avoid freeze-thaw cycles.