To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cbl proto-oncogene B, E3 Ubiquitin Protein Ligase
- Antigen Symbol:CBLB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:868
- Antigen Synonyms:EC 6.3.2.-|SH3-binding protein CBL-B|FLJ41152|DKFZp686J10223|Cas-Br-M (murine) ecotropic retroviral transforming sequence b|Cas-Br-M (murine) ectropic retroviral transforming sequence b|Signal transduction protein CBL-B|DKFZp779F1443|E3 ubiquitin-protein ligase CBL-B|RING finger protein 56|Casitas B-lineage lymphoma proto-oncogene b|EC 6.3.2|RNF56FLJ36865|DKFZp779A0729
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 932 and 982 of human Cas-Br-M (murine) Ecotropic Retroviral Transforming Sequence b using the numbering given in entry NP_733762.2 (GeneID 868).
- Purification:Immunogen affinity purified
- Cat. No.:102179-762
- Supplier no.:NBP1-49920
Specifications
About this item
The CBLB Antibody from Novus Biologicals is a rabbit polyclonal antibody to CBLB. This antibody reacts with human. The CBLB Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: CBLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human