Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:BTB (POZ) Domain Containing 15
- Antigen Symbol:BTBD15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:29068
- Antigen Synonyms:BTBD15|MGC60348|BTB (POZ) domain containing 15|zinc finger and BTB domain-containing protein 44|MGC88058|MGC57431|zinc finger and BTB domain containing 44|HSPC063|ZNF851MGC26123|Zinc finger protein 851|BTB/POZ domain-containing protein 15
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 300 and 350 of human Zinc Finger and BTB Domain Containing 44 using the numbering given in entry BAI46608.1 (GeneID 29068).
- Purification:Immunogen affinity purified
- Cat. No.:102212-564
- Supplier no.:NBP1-78184
The BTBD15 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BTBD15. This antibody reacts with human. The BTBD15 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation.
Type: Primary
Antigen: BTBD15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human