Order Entry
United States
ContactUsLinkComponent
Anti-B4GALT6 Rabbit Polyclonal Antibody
Anti-B4GALT6 Rabbit Polyclonal Antibody
Catalog # 102198-354
Supplier:  Novus Biologicals
Anti-B4GALT6 Rabbit Polyclonal Antibody
Catalog # 102198-354
Supplier:  Novus Biologicals
Supplier Number:  NBP1-69019

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Beta-1,4-galactosyltransferase 6
  • Antigen Symbol:
    B4GALT6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    9331
  • Antigen Synonyms:
    UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase|beta-1,4-galactosyltransferase 6|polypeptide 6|Beta-1,4-GalTase 6|beta4GalT-VI|UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6|B4Gal-T6|UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6|UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase|Beta4Gal-T6|EC 2.4.1.-
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to B4galt6 (UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 6) The peptide sequence was selected from the C terminal of B4galt6 (NP_062711). Peptide sequence GGEDDDLWNRVHYAGYNVTRPEGDLGKYISIPHHHRGEVQFLGRY
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102198-354
  • Supplier no.:
    NBP1-69019

Specifications

About this item

The B4GALT6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to B4GALT6. This antibody reacts with mouse. The B4GALT6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: B4GALT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse