Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Beta-1,3-Galactosyltransferase 2
- Antigen Symbol:B3GALT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:8707
- Antigen Synonyms:beta-1,3-galactosyltransferase 2|Beta3GalT2|beta-3-galt2|UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase|Beta-1,3-GalTase 2|UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2|beta3Gal-T2|EC 2.4.1.-|EC 2.4.1|GLCT2|polypeptide 2
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to B3galt2 (UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2) The peptide sequence was selected from the C terminal of B3galt2. Peptide sequence RVDPVPPPNEFVFNHWRVSYSSCKYSHLITSHQFQPSELIKYWNH
- Purification:Immunogen affinity purified
- Cat. No.:102198-344
- Supplier no.:NBP1-69014
Specifications
About this item
The B3GALT2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to B3GALT2. This antibody reacts with mouse. The B3GALT2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.
Type: Primary
Antigen: B3GALT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse