Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Mesencephalic astrocyte-derived neurotrophic Factor
- Antigen Symbol:ARMET
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:7873
- Antigen Synonyms:mutated in early stage tumors|Protein ARMET|arginine-rich|mesencephalic astrocyte-derived neurotrophic factor|MGC142148|ARPMGC142150|ARMETArginine-rich protein
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 12 amino acid synthetic peptide from near the amino terminus of human MANF. The immunogen is located within the first 50 amino acids of MANF.
- Purification:Immunogen affinity purified
- Cat. No.:102209-560
- Supplier no.:NBP1-76837
Specifications
About this item
The ARMET Antibody from Novus Biologicals is a rabbit polyclonal antibody to ARMET. This antibody reacts with human, mouse, rat. The ARMET Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ARMET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat