Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Anaphase Promoting Complex Subunit 5
- Antigen Symbol:Apc5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:51433
- Antigen Synonyms:anaphase promoting complex subunit 5|anaphase-promoting complex subunit 5|APC5Cyclosome subunit 5
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 705 and the C-terminus (residue 755) of human anaphase promoting complex subunit 5 using the numbering given in entry NP_057321.2 (GeneID 51433).
- Purification:Immunogen affinity purified
- Cat. No.:102109-696
- Supplier no.:NB100-55319
The Apc5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Apc5. This antibody reacts with human, mouse. The Apc5 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: Apc5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse