Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Aldosterone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:0.5 ml
- Storage Buffer:PBS (pH 7.2)
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:5.0 mg/ml
- Immunogen:Aldosterone-3-CMO-BSA
- Purification:Protein G purified
- Cat. No.:102114-020
- Supplier no.:NB100-64658
Specifications
About this item
The Aldosterone Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aldosterone. This antibody reacts with multi-species. The Aldosterone Antibody has been validated for the following applications: ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Aldosterone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Multi-Species