Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:AF4
- Antigen Symbol:AF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4299
- Antigen Synonyms:MLLT2MGC134969|AF4/FMR2 family member 1|Proto-oncogene AF4|PBM1|AF4myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog)|Protein AF-4|pre-B-cell monocytic leukemia partner 1|translocated to|myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog|ALL1-fused gene from chromosome 4 protein|member 1|FEL|Protein FEL|Drosophila)|2|AF4/FMR2 family|AF-4
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human proto-oncogene AF4 using the numbering given in entry NP_005926.1 (GeneID 4299).
- Purification:Immunogen affinity purified
- Cat. No.:102162-598
- Supplier no.:NBP1-28728
The AF4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AF4. This antibody reacts with human. The AF4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation (Negative).
Type: Primary
Antigen: AF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human