Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Adenylate Cyclase 2
- Antigen Symbol:ADCY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:108
- Antigen Synonyms:MGC133314|FLJ45092|Adenylate cyclase type II|adenylate cyclase 2 (brain)|AC2|type II adenylate cyclase|adenylate cyclase type 2|Adenylyl cyclase 2|KIAA1060FLJ16822|HBAC2|EC 4.6.1.1|3'-5'-cyclic AMP synthetase|ATP pyrophosphate-lyase 2|adenylate cyclase II
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.33 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Adenylate cyclase 2. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102167-296
- Supplier no.:NBP1-33659
Specifications
About this item
The Adenylate Cyclase 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Adenylate Cyclase 2. This antibody reacts with human. The Adenylate Cyclase 2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Adenylate Cyclase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human