Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:ATP Binding Cassette, Subfamily B, Member 9
- Antigen Symbol:ABCB9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:23457
- Antigen Synonyms:ATP-binding cassette transporter 9|EC 3.6.3.44|EC 3.6.3|TAP-like protein|KIAA1520|EST122234|ATP-binding cassette|hABCB9|sub-family B (MDR/TAP)|ATP-binding cassette sub-family B member 9|TAPL|ABC transporter 9 protein|member 9
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to ABCB9(ATP-binding cassette, sub-family B (MDR/TAP), member 9) The peptide sequence was selected from the N terminal of ABCB9. Peptide sequence RLWKAVVVTLAFMSVDICVTTAIYVFSHLDRSLLEDIRHFNIFDSVLDLW.
- Purification:Immunogen affinity purified
- Cat. No.:102199-068
- Supplier no.:NBP1-69513
Specifications
About this item
The ABCB9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ABCB9. This antibody reacts with human. The ABCB9 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ABCB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human