Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:200 kDa Neurofilament Heavy
- Antigen Symbol:NF-H
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken,Rat
- Western Blot:Yes
- Size:100 μl
- Gene ID:4744
- Antigen Synonyms:heavy polypeptide|Neurofilament Heavy (200kDa)|Neurofilament triplet H protein|KIAA0845|neurofilament heavy polypeptide|NF-H|neurofilament|NFH|NF200|200 kDa neurofilament protein|heavy polypeptide 200kDa
- Storage Buffer:Whole antisera
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Purified rat Neurofilament Heavy construct containing most of the tandem KSP repeats [UniProt# P16884]
- Purification:Unpurified
- Cat. No.:102156-720
- Supplier no.:NBP1-05215
Specifications
About this item
The NF-H Antibody from Novus Biologicals is a rabbit polyclonal antibody to NF-H. This antibody reacts with rat, chicken. The NF-H Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: NF-H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Chicken