To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:X-ray Repair Complementing Defective Repair in Chinese cells 1
- Antigen Symbol:XRCC1
- Clonality:Monoclonal
- Clone:2F8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7515
- Antigen Synonyms:complementing defective|RCC|X-ray-repair|X-ray repair complementing defective repair in Chinese hamster cells 1|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1|repair in Chinese hamster
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human XRCC1 (NP_006288) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102231-296
- Supplier no.:NBP2-02280
The XRCC1 Antibody (2F8) from Novus Biologicals is a mouse monoclonal antibody to XRCC1. This antibody reacts with human. The XRCC1 Antibody (2F8) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: XRCC1
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human