Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen Symbol:UCH-L1/PGP9.5
- Clonality:Monoclonal
- Clone:10A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Gene ID:7345
- Antigen Synonyms:Neuron cytoplasmic protein 9.5|EC 3.4.19.12|PGP9.5|UCHL1|PGP9.5Uch-L1|ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|ubiquitin carboxyl-terminal hydrolase isozyme L1|ubiquitin C-terminal hydrolase|PGP95|EC 6.-|PARK5
- Storage Buffer:Tissue culture supernatant
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:4.1 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Purification:Tissue culture supernatant
- Cat. No.:102106-374
- Supplier no.:NB100-1926
Specifications
About this item
The UCH-L1 / PGP9.5 Antibody (10A1) from Novus Biologicals is a mouse monoclonal antibody to UCH-L1 / PGP9.5. This antibody reacts with human, mouse, rat. The UCH-L1 / PGP9.5 Antibody (10A1) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: UCH-L1/PGP9.5
Clonality: Monoclonal
Clone: 10A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat