Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:ZFP36 Ring Finger Protein
- Antigen Symbol:TTP
- Clonality:Monoclonal
- Clone:8B5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7538
- Antigen Synonyms:EZNFTranscription factor 17|TCF17|zinc finger protein 354A|KID-1|KID1TCF-17|HKL1Zinc finger protein eZNF
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human ZFP36 (NP_003398) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102230-946
- Supplier no.:NBP2-02105
The TTP Antibody (8B5) from Novus Biologicals is a mouse monoclonal antibody to TTP. This antibody reacts with human, monkey. The TTP Antibody (8B5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TTP
Clonality: Monoclonal
Clone: 8B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey