Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Thyroid Stimulating Hormone Beta
- Antigen Symbol:TSH beta
- Clonality:Monoclonal
- Clone:110000
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:7252
- Antigen Synonyms:TSH-BETA|thyrotropin subunit beta|thyroid stimulating hormone|thyrotropin beta chain|beta|thyroid-stimulating hormone subunit beta|TSH-B|thyrotropin beta subunit
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Full length native protein (purified) from human pituitary gland.
- Purification:Protein A purified
- Cat. No.:102125-850
- Supplier no.:NB110-8274
Specifications
About this item
The TSH beta Antibody (11E4) from Novus Biologicals is a mouse monoclonal antibody to TSH beta. This antibody reacts with human. The TSH beta Antibody (11E4) has been validated for the following applications: Sandwich ELISA.
Type: Primary
Antigen: TSH beta
Clonality: Monoclonal
Clone: 110000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human