To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:TGF-beta RIII
- Antigen Symbol:TGFRB3
- Clonality:Monoclonal
- Clone:MM0057-5G9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:7049
- Antigen Synonyms:betaglycan|300kDa)|betaglycan proteoglycan|Transforming growth factor beta receptor III|beta receptor III|transforming growth factor|TGFR-3|transforming growth factor beta receptor type 3|BGCAN|TGF-beta receptor type III|TGF-beta receptor type 3|beta receptor III (betaglycan
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human TGF beta Receptor III
- Purification:Protein G purified
- Cat. No.:102250-576
- Supplier no.:NBP2-11920
Specifications
About this item
The TGF-beta RIII Antibody (MM0057-5G9) from Novus Biologicals is a mouse monoclonal antibody to TGF-beta RIII. This antibody reacts with human. The TGF-beta RIII Antibody (MM0057-5G9) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TGF-beta RIII
Clonality: Monoclonal
Clone: MM0057-5G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human