Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Sirtuin 1
- Antigen Symbol:SIRT1
- Clonality:Monoclonal
- Clone:1F3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:23411
- Antigen Synonyms:S. cerevisiae|EC 3.5.1|sirtuin 1|homolog) 1
- Storage Buffer:PBS
- Storage Temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Purified recombinant fragment of human SIRT1 (amino acids: 265-452) expressed in E. coli. [UniProt# Q96EB6]
- Purification:Ammonium sulfate precipitation
- Cat. No.:102182-000
- Supplier no.:NBP1-51641
Specifications
About this item
The Sirtuin 1 / SIRT1 Antibody (1F3) from Novus Biologicals is a mouse monoclonal antibody to Sirtuin 1 / SIRT1. This antibody reacts with human, mouse, rat, primate. The Sirtuin 1 / SIRT1 Antibody (1F3) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000
Type: Primary
Antigen: SIRT1
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate