Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Serine Dehydratase
- Antigen Symbol:SDH
- Clonality:Monoclonal
- Clone:3D3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10993
- Antigen Synonyms:L-serine deaminase|SDHL-serine dehydratase|L-serine dehydratase/L-threonine deaminase|L-threonine dehydratase|EC 4.3.1.17|L-serine ammonia-lyase|TDH|serine dehydratase|EC 4.3.1.19
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human SDS(NP_006834) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102229-748
- Supplier no.:NBP2-01506
The Serine Dehydratase Antibody (3D3) from Novus Biologicals is a mouse monoclonal antibody to Serine Dehydratase. This antibody reacts with human. The Serine Dehydratase Antibody (3D3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Serine Dehydratase
Clonality: Monoclonal
Clone: 3D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human