Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:SERCA2 ATPase
- Antigen Symbol:SERCA2
- Clonality:Monoclonal
- Clone:2A7-A1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Pig,Rabbit,Amphibian,Sheep,Human,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:488
- Antigen Synonyms:MGC45367|cardiac muscle|DAR|EC 3.6.3.8|Ca++ transporting|SR Ca(2+)-ATPase 2|DD|cardiac Ca2+ ATPase|Ca++ dependent|Calcium pump 2|SERCA2DKFZp686P0211|cardiac muscle-2|slow-twitch|slow twitch 2|EC 3.6.3|Endoplasmic reticulum class 1/2 Ca(2+) ATPase|Calcium-transporting ATPase sarcoplasmic reticulum type|ATP2BFLJ20293|sarcoplasmic/endoplasmic reticulum calcium ATPase 2|FLJ38063|slow twitch skeletalmuscle isoform|ATPase
- Storage Buffer:Ascites diluted with PBS
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Purified canine cardiac sarcoplasmic reticulum vesicles.
- Purification:Unpurified
- Cat. No.:102130-844
- Supplier no.:NB300-581
Specifications
About this item
The SERCA2 ATPase Antibody (2A7-A1) from Novus Biologicals is a mouse monoclonal antibody to SERCA2 ATPase. This antibody reacts with human, mouse, rat, porcine, amphibian, canine, guinea pig, rabbit, sheep. The SERCA2 ATPase Antibody (2A7-A1) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence, Immunocytochemistry.
Type: Primary
Antigen: SERCA2 ATPase
Clonality: Monoclonal
Clone: 2A7-A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Porcine, Amphibian, Canine, Guinea Pig, Rabbit, Sheep