Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:RNase L
- Antigen Symbol:RNASEL
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Hamster,Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:6041
- Antigen Synonyms:Ribonuclease 4|MGC104972|RNase L|2-5A-dependent ribonuclease|HPC1|PRCA1|ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent)|2-5A-dependent RNase|interferon-induced 2-5A-dependent RNase|Ribonuclease L|2'-5'-oligoisoadenylate synthetase-dependent|MGC133329|EC 3.1.26.-|prostate cancer 1|RNS4DKFZp781D08126
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823]
- Purification:Protein G purified
- Cat. No.:102108-574
- Supplier no.:NB100-351B
Specifications
About this item
The RNase L Antibody (2E9) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to RNase L. This antibody reacts with human, hamster, primate, mouse (negative), rat (negative). The RNase L Antibody (2E9) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: RNase L
Clonality: Monoclonal
Clone: 2E9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Hamster, Primate, Mouse (Negative), Rat (Negative)