Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:OATP2/8
- Clonality:Monoclonal
- Clone:MDQ
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:500 μl
- Epitope:The exact epitope within the immunizing peptide sequence is unknown.
- Gene ID:10599
- Antigen Synonyms:OATP-C|HBLRR|SLCO1B1 solute carrier organic anion transporter family|member 1B1|LST-1|OATP-8|SLC21A8|OATP1B3|OATP1B1|LST-2|OATP2|SLC21A6|OATP8|LST3|OATPC|LST-3TM13|LST1
- Storage Buffer:Tissue culture supernatant
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to residues M(1) D Q N Q H L N K T A E A Q P S E N K K T R Y(23) C of human OATP2.
- Purification:Unpurified
- Cat. No.:102116-404
- Supplier no.:NB100-74482
The OATP2 / 8 Antibody (MDQ) from Novus Biologicals is a mouse monoclonal antibody to OATP2 / 8. This antibody reacts with human. The OATP2 / 8 Antibody (MDQ) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: OATP2/8
Clonality: Monoclonal
Clone: MDQ
Conjugation: Unconjugated
Epitope: The exact epitope within the immunizing peptide sequence is unknown.
Host: Mouse
Isotype: IgG1
Reactivity: Human