Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Neurotrophin 4
- Antigen Symbol:NT-4
- Clonality:Monoclonal
- Clone:2E8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4909
- Antigen Synonyms:neurotrophin-4|NT-5|NT-4/5|Neutrophic factor 4|neurotrophic factor 5|neurotrophin 5 (neurotrophin 4/5)|neurotrophin 4|neurotrophic factor 4|NT-4|neurotrophin-5|NTF5NT5|GLC10|GLC1O|NT4
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Recombinant protein expressed in E.coli corresponding to amino acids 81-210 of human NT4
- Purification:Immunogen affinity purified
- Cat. No.:102178-028
- Supplier no.:NBP1-47897
Specifications
About this item
The NT-4 Antibody (2E8) from Novus Biologicals is a mouse monoclonal antibody to NT-4. This antibody reacts with human. The NT-4 Antibody (2E8) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NT-4
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human