Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Netrin-4
- Antigen Symbol:NTN4
- Clonality:Monoclonal
- Clone:Netry-2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:59277
- Antigen Synonyms:netrin 4|hepar-derived netrin-like protein|NTN4|beta-netrin|PRO3091
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant human netrin-4.
- Purification:Protein G purified
- Cat. No.:102222-160
- Supplier no.:NBP1-97616
The Netrin-4 Antibody (Netry-2) from Novus Biologicals is a mouse monoclonal antibody to Netrin-4. This antibody reacts with human, mouse. The Netrin-4 Antibody (Netry-2) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.
Type: Primary
Antigen: Netrin-4
Clonality: Monoclonal
Clone: Netry-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse