Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Natural Cytotoxicity Triggering Receptor 1
- Antigen Symbol:NKp46/NCR1
- Clonality:Monoclonal
- Clone:MM0491-8F24
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Lyophilized
- Gene ID:9437
- Antigen Synonyms:NK-p46)|NKP46FLJ99094|CD335|NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor|hNKp46|LY94lymphocyte antigen 94 homolog (activating NK-receptor|CD335 antigen|natural cytotoxicity triggering receptor 1|NK cell-activating receptor|Lymphocyte antigen 94 homolog|Natural killer cell p46-related protein|NKp46
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:Human NCR1
- Tested Applications:CyTOF-ready
- Purification:Protein G purified
- Cat. No.:102250-376
- Supplier no.:NBP2-11820
Specifications
About this item
The NKp46 / NCR1 Antibody (MM0491-8F24) from Novus Biologicals is a mouse monoclonal antibody to NKp46 / NCR1. This antibody reacts with human. The NKp46 / NCR1 Antibody (MM0491-8F24) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
Type: Primary
Antigen: NKp46/NCR1
Clonality: Monoclonal
Clone: MM0491-8F24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human