Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Kelch-like 11
- Antigen Symbol:KLHL11
- Clonality:Monoclonal
- Clone:1B9C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:55175
- Antigen Synonyms:kelch-like 11 (Drosophila)|FLJ10572|kelch-like protein 11
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Purified recombinant fragment of human KLHL11 expressed in E. Coli.
- Purification:Unpurified
- Cat. No.:102181-970
- Supplier no.:NBP1-51626
Mouse Monoclonal KLHL11 Antibody (1B9C1). Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000
Type: Primary
Antigen: KLHL11
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human