Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin alpha 6
- Antigen Symbol:ITGA6
- Clonality:Monoclonal
- Clone:450-30A
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:3655
- Antigen Synonyms:CD49f antigen|CD49 antigen-like family member F|integrin alpha-6|DKFZp686J01244|alpha 6|integrin alpha6B|CD49f|FLJ18737|VLA-6|integrin|ITGA6B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:alpha 6 beta 4 integrin purified from A431 cells
- Purification:Protein G purified
- Cat. No.:102114-286
- Supplier no.:NB100-64908
Specifications
About this item
Mouse Monoclonal Integrin alpha 6 Antibody (450-30A). Tested Applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Human.
Type: Primary
Antigen: Integrin alpha 6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human