Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 1 Beta
- Antigen Symbol:IL-1 beta/IL-1F2
- Clonality:Monoclonal
- Clone:60000000000
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:0.2 mg
- Gene ID:3553
- Antigen Synonyms:catabolin|IL-1 beta|preinterleukin 1 beta|IL-1B|pro-interleukin-1-beta|IL1-BETA|interleukin-1 beta|beta|IL1F2IL-1 beta|IL-1|interleukin 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Recombinant beta human Interleukin-1
- Purification:Protein A purified
- Cat. No.:102128-876
- Supplier no.:NB120-8319
Specifications
About this item
Mouse Monoclonal IL1 beta Antibody (6E10). Tested Applications: Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Sandwich ELISA. Tested Reactivity: Human.
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human