Order Entry
United States
ContactUsLinkComponent
Anti-Glutathione Synthetase Mouse Monoclonal Antibody [clone: 1B8]
Anti-Glutathione Synthetase Mouse Monoclonal Antibody [clone: 1B8]
Catalog # 102233-438
Supplier:  Novus Biologicals
Anti-Glutathione Synthetase Mouse Monoclonal Antibody [clone: 1B8]
Catalog # 102233-438
Supplier:  Novus Biologicals
Supplier Number:  NBP2-03351
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Glutathione Synthetase
  • Antigen Symbol:
    GSS
  • Clonality:
    Monoclonal
  • Clone:
    1B8
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    2937
  • Antigen Synonyms:
    GSH synthetase|GSH-S|MGC14098|GSHS|glutathione synthetase|Glutathione synthase|EC 6.3.2.3
  • Storage Buffer:
    PBS (pH 7.3), 1.0% BSA and 50% Glycerol
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Full length human recombinant protein of human GSS (NP_000169) produced in HEK293T cell.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102233-438
  • Supplier no.:
    NBP2-03351

Specifications

About this item

The Glutathione Synthetase Antibody (1B8) from Novus Biologicals is a mouse monoclonal antibody to Glutathione Synthetase. This antibody reacts with human. The Glutathione Synthetase Antibody (1B8) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Glutathione Synthetase
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human