Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:GATA binding Protein 4
- Antigen Symbol:GATA-4
- Clonality:Monoclonal
- Clone:9F9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:2626
- Antigen Synonyms:GATA binding protein 4|GATA-binding factor 4|MGC126629|GATA-binding protein 4|transcription factor GATA-4
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Recombinant fragment expressed in E.coli corresponding to amino acids 296-442of human GATA4
- Purification:Immunogen affinity purified
- Cat. No.:102177-764
- Supplier no.:NBP1-47765
Specifications
About this item
The GATA-4 Antibody (9F9) from Novus Biologicals is a mouse monoclonal antibody to GATA-4. This antibody reacts with human. The GATA-4 Antibody (9F9) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: GATA-4
Clonality: Monoclonal
Clone: 9F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human