The GAPDH Antibody (NB615) from Novus Biologicals is a mouse monoclonal antibody to GAPDH. This antibody reacts with human, mouse, rat, porcine, bovine, canine, chicken, rabbit. The GAPDH Antibody (NB615) has been validated for the following applications: Western Blot, Simple Western, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: NB615
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Rabbit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.