To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:gamma H2AX
- Antigen Symbol:H2AXG
- Clonality:Monoclonal
- Clone:3F2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine,Mouse
- Western Blot:Yes
- Size:100 ug
- Environmentally Preferable:
- Gene ID:3014
- Antigen Synonyms:member X|H2AX histone|H2a/x|H2A.X|H2AXhistone H2A.x|H2A histone family|YH2AX
- Modification:p Ser140
- Storage Buffer:PBS with 1 mg/ml BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide sequence surrounding phosphorylated Ser140
- Purification:Protein G purified
- Cat. No.:102116-356
- Supplier no.:NB100-74435
The gamma H2AX [p Ser140] Antibody (3F2) from Novus Biologicals is a mouse monoclonal antibody to gamma H2AX. This antibody reacts with human, mouse, bovine. The gamma H2AX [p Ser140] Antibody (3F2) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: gamma H2AX
Clonality: Monoclonal
Clone: 3F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Bovine