Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:FK506 Binding Protein 12
- Antigen Symbol:FKBP12
- Clonality:Monoclonal
- Clone:3B3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:2280
- Antigen Synonyms:FK506 binding protein 1A|FKBP12C|FKBP1|EC 5.2.1.8|12kDa|12 kDa FKBP|FKBP-1A|PPIase FKBP1A|protein kinase C inhibitor 2|peptidyl-prolyl cis-trans isomerase FKBP1A|12-kD|FKBP12FKBP12-Exip3|FKBP-12T-cell|FK506-binding protein 1A|rotamase
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human FKBP1A (NP_000792) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102230-922
- Supplier no.:NBP2-02093
Specifications
About this item
The FKBP12 Antibody (3B3) from Novus Biologicals is a mouse monoclonal antibody to FKBP12. This antibody reacts with human. The FKBP12 Antibody (3B3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FKBP12
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human