Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Ferritin Heavy Chain
- Antigen Symbol:FTH1
- Clonality:Monoclonal
- Clone:3F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 ml
- Gene ID:2495
- Antigen Synonyms:FHC|proliferation-inducing protein 15|PLIF|ferritin|PIG15|ferritin heavy chain|placenta immunoregulatory factor|Cell proliferation-inducing gene 15 protein|Ferritin H subunit|heavy polypeptide 1|FTHEC 1.16.3.1|FTHL6MGC104426|apoferritin
- Storage Buffer:HEPES, 0.01% BSA, 0.15M NaCl and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant human protein purified from E. coli (HIS-Ferritin H chain).
- Purification:Ammonium sulfate precipitation
- Cat. No.:102249-856
- Supplier no.:NBP2-11536
Specifications
About this item
The Ferritin Heavy Chain Antibody (3F8) from Novus Biologicals is a mouse monoclonal antibody to Ferritin Heavy Chain. This antibody reacts with human. The Ferritin Heavy Chain Antibody (3F8) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: Ferritin Heavy Chain
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 Kappa
Reactivity: Human